Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HSPA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016299.3 | NP_057383.2 | 12 | 1473 | ||
GenBank mRNA | AF112210.1 | - | 12 | 1292 | ||
AF143723.1 | - | 12 | 1282 | |||
AK225265.1 | - | 12 | 1317 | |||
AK225651.1 | - | 12 | 1317 | |||
AK226065.1 | - | 5 | 4146 | |||
AK292323.1 | - | 12 | 1281 | |||
BC119690.1 | - | 12 | 1292 | 40116697 |