Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LUC7L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006107.3 | NP_006098.2 | 1 | 186 | ||
NM_016424.4 | NP_057508.2 | 1 | 186 | |||
XM_005257448.1 | XP_005257505.1 | 1 | 154 | |||
XM_005257449.2 | XP_005257506.1 | 1 | 154 | |||
XM_005257452.1 | XP_005257509.1 | 1 | 154 | |||
GenBank mRNA | AK001925.1 | - | 1 | 120 | ||
AK023672.1 | - | 1 | 148 | |||
AK225980.1 | - | 1 | 154 | |||
AK290540.1 | - | 1 | 153 | |||
AK295983.1 | - | 1 | 122 | |||
BC002925.2 | - | 1 | 127 | |||
BC047043.1 | - | 1 | 135 | |||
BC056409.1 | - | 1 | 133 | |||
DA231851.1 | - | 1 | 186 | |||
DQ013876.1 | - | 1 | 122 |