Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SERPINE2 SIRNA Assays ›
          Gene Symbol:
          SERPINE2
          Gene Name:
          serpin family E member 2
          Gene Aliases:
          GDN, GDNPF, PI-7, PI7, PN-1, PN1, PNI
          Chromosome Location:
          Chr.2: 223975048 - 224039319 on Build GRCh38
          UniGene ID:
          Hs.38449
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS182297
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SERPINE2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136528.1 NP_001130000.1 3 592
          NM_001136530.1 NP_001130002.1 3 522
          NM_006216.3 NP_006207.1 3 592
          NR_073116.1 4 1010
          XM_005246641.2 XP_005246698.1 3 681
          XM_017004329.1 XP_016859818.1 3 573
          XM_017004330.1 XP_016859819.1 3 410
          XM_017004331.1 XP_016859820.1 3 573
          XM_017004332.1 XP_016859821.1 3 410
          GenBank mRNA A03911.1 - 2 373
          AK225823.1 - 3 560
          AK295564.1 - 3 490
          AK297588.1 - 3 397
          AK312499.1 - 3 606
          BC015663.2 - 3 549 4578406
          BC042628.1 - 4 1009 4824856
          DC304261.1 - 3 522
          M17783.1 - 2 349

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          embryo implantation
          blood coagulation
          negative regulation of cell proliferation
          response to radiation
          negative regulation of plasminogen activation
          negative regulation of sodium ion transport
          negative regulation of endopeptidase activity
          negative regulation of protein processing
          positive regulation of neuron projection development
          negative regulation of phosphatidylinositol 3-kinase signaling
          cerebellar granular layer morphogenesis
          negative regulation of blood coagulation
          negative regulation of cell growth
          regulation of cell migration
          secretion by cell
          secretory granule organization
          negative regulation of protein catabolic process
          mating plug formation
          negative regulation of proteolysis
          negative regulation of smoothened signaling pathway
          regulation of timing of cell differentiation
          positive regulation of astrocyte differentiation
          detection of mechanical stimulus involved in sensory perception
          regulation of synaptic transmission, glutamatergic
          long-term synaptic potentiation
          innervation
          seminal vesicle epithelium development
          dense core granule biogenesis
          negative regulation of platelet aggregation
          serine-type endopeptidase inhibitor activity
          receptor binding
          protein binding
          glycosaminoglycan binding
          heparin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jct7h:80/100.66.72.173:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0