Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADAM22 SIRNA Assays ›
Gene Symbol:
ADAM22
Gene Name:
ADAM metallopeptidase domain 22
Gene Aliases:
ADAM 22, MDC2
Chromosome Location:
Chr.7: 87934143 - 88202889 on Build GRCh38
UniGene ID:
Hs.256398
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS182378
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADAM22
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324417.1
NP_001311346.1
24
2446
NM_001324418.1
NP_001311347.1
24
2449
NM_001324419.1
NP_001311348.1
24
2446
NM_001324420.1
NP_001311349.1
24
2449
NM_001324421.1
NP_001311350.1
24
2449
NM_004194.4
NP_004185.1
24
2449
NM_016351.5
NP_057435.2
24
2449
NM_021721.4
NP_068367.1
24
2449
NM_021722.5
NP_068368.2
24
2449
NM_021723.4
NP_068369.1
24
2449
XM_005250445.3
XP_005250502.1
23
2297
XM_006716028.3
XP_006716091.1
23
2297
XM_006716029.3
XP_006716092.1
23
2297
XM_011516318.1
XP_011514620.1
24
2341
XM_011516319.2
XP_011514621.1
24
2425
XM_011516320.2
XP_011514622.1
23
2297
XM_011516321.2
XP_011514623.1
23
2297
XM_011516322.1
XP_011514624.1
24
2341
XM_011516323.1
XP_011514625.1
24
2341
XM_011516324.1
XP_011514626.1
24
2013
XM_017012329.1
XP_016867818.1
25
2392
XM_017012330.1
XP_016867819.1
25
2392
XM_017012331.1
XP_016867820.1
24
2422
XM_017012332.1
XP_016867821.1
24
2341
XM_017012333.1
XP_016867822.1
24
2341
XM_017012334.1
XP_016867823.1
25
2392
XM_017012335.1
XP_016867824.1
25
2392
XM_017012336.1
XP_016867825.1
24
2341
XM_017012337.1
XP_016867826.1
25
2392
XM_017012338.1
XP_016867827.1
24
2420
XM_017012339.1
XP_016867828.1
25
2392
XM_017012340.1
XP_016867829.1
13
1072
XM_017012341.1
XP_016867830.1
23
2069
GenBank mRNA
AB009671.1
-
24
2247
AF073291.2
-
24
2172
AF155381.1
-
24
2172
AF155382.1
-
24
2172
AF158637.1
-
23
2009
AK125432.1
-
23
2312
AK291010.1
-
24
2296
BC125118.1
-
24
2203
40118904
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
cell adhesion
negative regulation of cell adhesion
central nervous system development
adult locomotory behavior
myelination in peripheral nervous system
metalloendopeptidase activity
integrin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline