Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIF1A SIRNA Assays ›
Gene Symbol:
KIF1A
Gene Name:
kinesin family member 1A
Gene Aliases:
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location:
Chr.2: 240713764 - 240821025 on Build GRCh38
UniGene ID:
Hs.516802
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS182649
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIF1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001244008.1
NP_001230937.1
31
3488
NM_001320705.1
NP_001307634.1
30
3212
NM_004321.6
NP_004312.2
28
2998
XM_005247022.1
XP_005247079.1
31
3399
XM_005247024.1
XP_005247081.1
30
3372
XM_005247027.1
XP_005247084.1
29
3096
XM_005247028.2
XP_005247085.1
29
3099
XM_006712605.1
XP_006712668.1
30
3372
XM_011511364.1
XP_011509666.1
31
3411
XM_011511365.1
XP_011509667.1
30
3123
XM_011511366.2
XP_011509668.1
21
2471
XM_011511367.1
XP_011509669.1
21
2401
XM_017004383.1
XP_016859872.1
31
3474
XM_017004384.1
XP_016859873.1
31
3486
XM_017004385.1
XP_016859874.1
31
3474
XM_017004386.1
XP_016859875.1
31
3474
XM_017004387.1
XP_016859876.1
30
3198
XM_017004388.1
XP_016859877.1
30
3198
XM_017004389.1
XP_016859878.1
30
3198
XM_017004390.1
XP_016859879.1
29
3096
XM_017004391.1
XP_016859880.1
30
3253
GenBank mRNA
AB209138.1
-
4
297
AB210029.1
-
31
3488
AB290172.1
-
31
3488
BC111780.1
-
28
2966
40037563
BC111799.1
-
28
2966
40037561
KJ534875.1
-
28
2938
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
anterograde axonal transport
motor activity
microtubule motor activity
ATP binding
microtubule binding
ATPase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline