Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GOLPH3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018178.5 | NP_060648.2 | 2 | 287 | ||
XM_006711428.1 | XP_006711491.1 | 2 | 214 | |||
GenBank mRNA | AJ296153.1 | - | 2 | 115 | ||
AK300635.1 | - | 2 | 245 | |||
AK315594.1 | - | 2 | 254 | |||
BC013870.2 | - | 1 | 86 | 3837466 | ||
DA513375.1 | - | 2 | 287 |