Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206994.1 | NP_001193923.1 | 7 | 783 | ||
NM_001206995.1 | NP_001193924.1 | 7 | 887 | |||
NM_001206996.1 | NP_001193925.1 | 7 | 937 | |||
NM_020416.3 | NP_065149.2 | 6 | 1075 | |||
NM_181876.2 | NP_870991.1 | 6 | 729 | |||
XM_005247979.4 | XP_005248036.1 | 2 | 920 | |||
XM_011513495.1 | XP_011511797.1 | 6 | 824 | |||
XM_011513496.1 | XP_011511798.1 | 6 | 839 | |||
GenBank mRNA | AB209183.1 | - | 5 | 4351 | ||
AF086924.1 | - | 6 | 759 | |||
AF145027.1 | - | 6 | 922 | |||
AK093115.1 | - | 6 | 729 | |||
AK122849.1 | - | 2 | 2736 | |||
AK296474.1 | - | 7 | 783 | |||
AK316003.1 | - | 8 | 897 | |||
BC032954.2 | - | 7 | 937 | 4823280 |