Search
Search

CDV3| Gene Symbol: CDV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282762.1 | NP_001269691.1 | 4, 5 | 975 | ||
| NM_001282763.1 | NP_001269692.1 | 4, 5 | 502 | |||
| NM_017548.4 | NP_060018.1 | 4, 5 | 1079 | |||
| XM_005247590.4 | XP_005247647.1 | 4, 5 | 1103 | |||
| GenBank mRNA | AF103803.1 | - | 4, 5 | 947 | ||
| AK025647.1 | - | 1, 2 | 98 | |||
| BG773216.1 | - | 4, 5 | 507 | |||
| DA949679.1 | - | 4, 5 | 485 | |||