Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UBQLN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304342.1 | NP_001291271.1 | 2 | 325 | ||
NM_020131.4 | NP_064516.2 | 2 | 325 | |||
XM_005245348.3 | XP_005245405.1 | 2 | 230 | |||
GenBank mRNA | AB209566.1 | - | 2 | 198 | ||
AF188240.1 | - | 2 | 479 | |||
AK302895.1 | - | 2 | 214 | |||
AK314413.1 | - | 2 | 213 | |||
BC018403.2 | - | 2 | 213 | 4214386 | ||
BC063841.1 | - | 2 | 225 | 6183942 | ||
HY061664.1 | - | 2 | 329 |