Search
Search

ISLR2| Gene Symbol: ISLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130136.1 | NP_001123608.1 | 4 | 1579 | ||
| NM_001130137.1 | NP_001123609.1 | 4 | 1315 | |||
| NM_001130138.1 | NP_001123610.1 | 4 | 1023 | |||
| NM_020851.2 | NP_065902.1 | 3 | 1148 | |||
| XM_011521840.2 | XP_011520142.1 | 4 | 1418 | |||
| XM_011521841.1 | XP_011520143.1 | 4 | 1101 | |||
| XM_017022446.1 | XP_016877935.1 | 3 | 1250 | |||
| GenBank mRNA | AB040898.2 | - | 4 | 1579 | ||
| AK092297.1 | - | 3 | 974 | |||
| AK092302.1 | - | 4 | 1595 | |||
| AK096602.1 | - | 2 | 926 | |||
| AK290467.1 | - | 3 | 1017 | |||
| AY358316.1 | - | 3 | 946 | |||
| BC028042.1 | - | 4 | 1311 | |||
| BC152429.1 | - | 4 | 1579 | 8862545 | ||