Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020933.4 | NP_065984.3 | 5, 6 | 696 | ||
NR_102435.1 | 5, 6 | 842 | ||||
GenBank mRNA | AB046808.1 | - | 4, 5 | 473 | ||
AF275255.1 | - | 5, 6 | 679 | |||
AF307097.1 | - | 3, 4 | 915 | |||
AK056570.1 | - | 5, 6 | 679 | |||
AK314560.1 | - | 5, 6 | 679 | |||
BC078154.1 | - | 5, 6 | 666 | 6060418 |