Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RFWD2 SIRNA Assays ›
Gene Symbol:
RFWD2
Gene Name:
ring finger and WD repeat domain 2
Gene Aliases:
COP1, RNF200
Chromosome Location:
Chr.1: 175944826 - 176207279 on Build GRCh38
UniGene ID:
Hs.523744
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS184830
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RFWD2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001740.3
NP_001001740.1
3
757
NM_001286644.1
NP_001273573.1
3
757
NM_022457.6
NP_071902.2
3
757
XM_005245447.2
XP_005245504.1
3
792
XM_005245448.2
XP_005245505.1
3
792
XM_006711487.2
XP_006711550.1
3
792
XM_011509881.1
XP_011508183.1
3
792
XM_017002059.1
XP_016857548.1
3
792
XM_017002060.1
XP_016857549.1
3
792
XM_017002061.1
XP_016857550.1
3
792
XM_017002062.1
XP_016857551.1
3
792
XM_017002063.1
XP_016857552.1
3
792
XM_017002064.1
XP_016857553.1
3
792
XM_017002065.1
XP_016857554.1
3
792
XM_017002066.1
XP_016857555.1
3
792
XM_017002067.1
XP_016857556.1
3
792
XM_017002068.1
XP_016857557.1
3
792
XM_017002071.1
XP_016857560.1
2
84
XM_017002073.1
XP_016857562.1
2
84
XM_017002075.1
XP_016857564.1
3
218
XM_017002078.1
XP_016857567.1
3
146
XM_017002079.1
XP_016857568.1
3
573
GenBank mRNA
AF508940.1
-
3
491
AF527539.1
-
3
754
AK001278.1
-
2
56
AK314750.1
-
3
689
AY509921.1
-
3
491
BC020845.1
-
3
664
BC039723.1
-
3
204
BC094728.1
-
3
745
5271629
KJ534928.1
-
3
491
KJ534929.1
-
3
491
KJ534930.1
-
3
491
KJ534931.1
-
3
491
KJ534932.1
-
3
491
KJ534992.1
-
3
491
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_006250116, XM_006250114, XM_006250113, XM_006250115.
This siRNA also targets the following transcripts in Mouse: NM_011931, AF151110, BC158002, BC082804.
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
response to ionizing radiation
protein ubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-xjq9k:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0