Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SIAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006610.1 | NP_001006611.1 | 2 | 943 | ||
NM_003031.3 | NP_003022.3 | 2 | 571 | |||
XM_006721246.1 | XP_006721309.1 | 3 | 747 | |||
XM_011523279.1 | XP_011521581.1 | 6 | 2087 | |||
GenBank mRNA | AB451253.1 | - | 1 | 454 | ||
AB451377.1 | - | 1 | 454 | |||
AK094663.1 | - | 1 | 1908 | |||
AK313861.1 | - | 2 | 957 | |||
BC035562.1 | - | 2 | 571 | 5224755 | ||
BC042550.1 | - | 2 | 913 | 5198176 | ||
BC044920.1 | - | 6 | 2050 | 5295925 | ||
BX647064.1 | - | 1 | 1515 | |||
EF026094.1 | - | 1 | 454 | |||
U63295.1 | - | 1 | 454 | |||
U76247.1 | - | 2 | 640 |
Gene Symbol: LOC100507577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040677.1 | 2 | 1521 | |||
GenBank mRNA | AK056051.1 | - | 2 | 1520 |