Search
Search

| Gene Symbol: PINK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032409.2 | NP_115785.1 | 8 | 1606 | ||
| GenBank mRNA | AB053323.1 | - | 8 | 1606 | ||
| AF316873.1 | - | 8 | 1582 | |||
| AK075225.1 | - | 4 | 605 | |||
| AK130746.1 | - | 5 | 2071 | |||
| BC009534.1 | - | 5 | 679 | |||
| BC028215.1 | - | 8 | 1521 | 5214483 | ||
| Gene Symbol: PINK1-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046507.1 | 1 | 1733 | |||
| GenBank mRNA | AK172749.1 | - | 1 | 818 | ||