Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PHACTR4 SIRNA Assays ›
          Gene Symbol:
          PHACTR4
          Gene Name:
          phosphatase and actin regulator 4
          Gene Aliases:
          PPP1R124
          Chromosome Location:
          Chr.1: 28369574 - 28500369 on Build GRCh38
          UniGene ID:
          Hs.225641
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS185958
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PHACTR4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001048183.1 NP_001041648.1 13 2354
          NM_023923.3 NP_076412.3 12 2337
          XM_005245970.2 XP_005246027.1 13 2361
          XM_017002157.1 XP_016857646.1 13 2361
          XM_017002158.1 XP_016857647.1 14 2271
          XM_017002159.1 XP_016857648.1 14 2520
          XM_017002160.1 XP_016857649.1 12 2337
          XM_017002161.1 XP_016857650.1 14 2351
          XM_017002162.1 XP_016857651.1 15 2413
          XM_017002163.1 XP_016857652.1 13 2966
          XM_017002164.1 XP_016857653.1 13 2351
          XM_017002165.1 XP_016857654.1 14 2324
          XM_017002166.1 XP_016857655.1 14 2351
          XM_017002167.1 XP_016857656.1 15 2413
          XM_017002168.1 XP_016857657.1 14 2324
          XM_017002169.1 XP_016857658.1 14 2254
          XM_017002170.1 XP_016857659.1 12 2395
          XM_017002171.1 XP_016857660.1 15 2488
          XM_017002172.1 XP_016857661.1 12 2172
          XM_017002173.1 XP_016857662.1 14 2254
          GenBank mRNA AF130081.1 - 7 981
          AK023233.1 - 7 1262
          AK024663.1 - 2 149
          AK025436.1 - 14 2341
          BC021247.2 - 12 2316 4577873
          BC029266.1 - 13 2213 4422413
          BC068508.1 - 15 2343 4825777
          CR749449.1 - 13 2333

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          neural crest cell migration
          neural tube closure
          Rho protein signal transduction
          actin cytoskeleton organization
          positive regulation of catalytic activity
          enteric nervous system development
          regulation of cell cycle
          closure of optic fissure
          negative regulation of integrin-mediated signaling pathway
          actin binding
          protein phosphatase 1 binding
          protein phosphatase type 1 activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0