Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SSX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021015.3 | NP_066295.3 | 2 | 90 | ||
NM_175723.1 | NP_783729.1 | 2 | 90 | |||
XM_011543949.2 | XP_011542251.1 | 2 | 125 | |||
XM_017029758.1 | XP_016885247.1 | 2 | 125 | |||
GenBank mRNA | BC016640.2 | - | 1 | 56 | 3923927 | |
BE896093.1 | - | 1 | 70 | |||
U90842.1 | - | 1 | 37 |