Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STAT3 SIRNA Assays ›
          Gene Symbol:
          STAT3
          Gene Name:
          signal transducer and activator of transcription 3
          Gene Aliases:
          ADMIO, ADMIO1, APRF, HIES
          Chromosome Location:
          Chr.17: 42313324 - 42388505 on Build GRCh38
          UniGene ID:
          Hs.463059
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS186130
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STAT3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003150.3 NP_003141.2 2 230
          NM_139276.2 NP_644805.1 2 252
          NM_213662.1 NP_998827.1 2 144
          XM_005257616.3 XP_005257673.2 2 243
          XM_005257617.3 XP_005257674.2 2 238
          XM_011525145.2 XP_011523447.1 2 243
          XM_011525146.2 XP_011523448.1 2 243
          XM_017024972.1 XP_016880461.1 2 238
          XM_017024973.1 XP_016880462.1 2 243
          XM_017024974.1 XP_016880463.1 2 238
          XM_017024975.1 XP_016880464.1 2 231
          XM_017024976.1 XP_016880465.1 2 231
          GenBank mRNA AB451232.1 - 1 12
          AJ012463.1 - 1 12
          AK291933.1 - 2 325
          AK301200.1 - 2 240
          BC000627.2 - 2 149 3347434
          BC014482.1 - 2 144 4909141
          BI461226.1 - 2 257
          L29277.1 - 2 232

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          temperature homeostasis
          eye photoreceptor cell differentiation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          protein import into nucleus
          movement of cell or subcellular component
          acute-phase response
          signal transduction
          JAK-STAT cascade
          nervous system development
          aging
          cell proliferation
          negative regulation of cell proliferation
          positive regulation of gene expression
          negative regulation of hydrogen peroxide biosynthetic process
          viral process
          phosphorylation
          cytokine-mediated signaling pathway
          sexual reproduction
          intracellular receptor signaling pathway
          response to estradiol
          cellular response to hormone stimulus
          leptin-mediated signaling pathway
          somatic stem cell population maintenance
          miRNA mediated inhibition of translation
          regulation of multicellular organism growth
          response to drug
          positive regulation of tyrosine phosphorylation of Stat3 protein
          glucose homeostasis
          eating behavior
          negative regulation of apoptotic process
          cellular response to leptin stimulus
          response to leptin
          response to ethanol
          positive regulation of Notch signaling pathway
          negative regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of mitochondrial membrane permeability
          astrocyte differentiation
          regulation of cell cycle
          radial glial cell differentiation
          regulation of feeding behavior
          growth hormone receptor signaling pathway
          JAK-STAT cascade involved in growth hormone signaling pathway
          interleukin-6-mediated signaling pathway
          cellular response to organic cyclic compound
          energy homeostasis
          negative regulation of neuron death
          positive regulation of growth factor dependent skeletal muscle satellite cell proliferation
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          positive regulation of metalloendopeptidase activity
          positive regulation of gene silencing by miRNA
          positive regulation of ATP biosynthetic process
          negative regulation of neuron migration
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II repressing transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          signal transducer activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          transcription factor binding
          protein kinase binding
          protein phosphatase binding
          chromatin DNA binding
          CCR5 chemokine receptor binding
          glucocorticoid receptor binding
          identical protein binding
          transcription regulatory region DNA binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gh6bl:80/100.66.76.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0