Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC1 SIRNA Assays ›
Gene Symbol:
TACC1
Gene Name:
transforming acidic coiled-coil containing protein 1
Gene Aliases:
Ga55
Chromosome Location:
Chr.8: 38728186 - 38853028 on Build GRCh38
UniGene ID:
Hs.279245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS186178
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122824.1
NP_001116296.1
8
1144
NM_001146216.2
NP_001139688.1
10
2320
NM_006283.2
NP_006274.2
10
2374
XM_005273625.4
XP_005273682.1
12
2524
XM_005273628.1
XP_005273685.1
11
2031
XM_005273629.1
XP_005273686.1
12
2044
XM_005273630.1
XP_005273687.1
11
2052
XM_011544631.1
XP_011542933.1
11
2410
XM_011544632.2
XP_011542934.1
13
2585
XM_011544633.1
XP_011542935.1
11
2323
XM_011544634.1
XP_011542936.1
10
2287
XM_011544635.1
XP_011542937.1
11
2114
XM_011544636.2
XP_011542938.1
8
770
XM_011544637.1
XP_011542939.1
8
1057
XM_011544638.2
XP_011542940.1
8
660
XM_017013766.1
XP_016869255.1
10
1995
XM_017013767.1
XP_016869256.1
11
2008
XM_017013768.1
XP_016869257.1
10
1908
XM_017013769.1
XP_016869258.1
11
1921
XM_017013770.1
XP_016869259.1
10
1944
XM_017013771.1
XP_016869260.1
10
2216
XM_017013772.1
XP_016869261.1
9
1908
XM_017013773.1
XP_016869262.1
9
2175
XM_017013774.1
XP_016869263.1
9
1860
XM_017013775.1
XP_016869264.1
10
1929
XM_017013776.1
XP_016869265.1
11
1921
XM_017013777.1
XP_016869266.1
9
2085
XM_017013778.1
XP_016869267.1
9
1784
GenBank mRNA
AB029026.1
-
9
941
AF049910.1
-
10
2315
AK294931.1
-
10
1888
AK295071.1
-
10
1922
AK304507.1
-
12
2120
AK304725.1
-
7
629
AK314620.1
-
10
2374
AY039239.1
-
9
684
AY139007.1
-
2
1019
AY177411.1
-
8
1085
BC041391.1
-
12
2484
5276767
BX647696.1
-
10
2319
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell cycle
cell proliferation
cerebral cortex development
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-pq62h:80/100.66.74.89:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0