Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TACC1 SIRNA Assays ›
          Gene Symbol:
          TACC1
          Gene Name:
          transforming acidic coiled-coil containing protein 1
          Gene Aliases:
          Ga55
          Chromosome Location:
          Chr.8: 38728186 - 38853028 on Build GRCh38
          UniGene ID:
          Hs.279245
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS186179
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TACC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122824.1 NP_001116296.1 8, 9 1278
          NM_001146216.2 NP_001139688.1 10, 11 2454
          NM_006283.2 NP_006274.2 10, 11 2508
          XM_005273625.4 XP_005273682.1 12, 13 2658
          XM_005273628.1 XP_005273685.1 11, 12 2165
          XM_005273629.1 XP_005273686.1 12, 13 2178
          XM_005273630.1 XP_005273687.1 11, 12 2186
          XM_011544631.1 XP_011542933.1 11, 12 2544
          XM_011544632.2 XP_011542934.1 13, 14 2719
          XM_011544633.1 XP_011542935.1 11, 12 2457
          XM_011544634.1 XP_011542936.1 10, 11 2421
          XM_011544635.1 XP_011542937.1 11, 12 2248
          XM_011544636.2 XP_011542938.1 8, 9 904
          XM_011544637.1 XP_011542939.1 8, 9 1191
          XM_011544638.2 XP_011542940.1 8, 9 794
          XM_017013766.1 XP_016869255.1 10, 11 2129
          XM_017013767.1 XP_016869256.1 11, 12 2142
          XM_017013768.1 XP_016869257.1 10, 11 2042
          XM_017013769.1 XP_016869258.1 11, 12 2055
          XM_017013770.1 XP_016869259.1 10, 11 2078
          XM_017013771.1 XP_016869260.1 10, 11 2350
          XM_017013772.1 XP_016869261.1 9, 10 2042
          XM_017013773.1 XP_016869262.1 9, 10 2309
          XM_017013774.1 XP_016869263.1 9, 10 1994
          XM_017013775.1 XP_016869264.1 10, 11 2063
          XM_017013776.1 XP_016869265.1 11, 12 2055
          XM_017013777.1 XP_016869266.1 9, 10 2219
          XM_017013778.1 XP_016869267.1 9, 10 1918
          GenBank mRNA AB029026.1 - 9, 10 1075
          AF049910.1 - 10, 11 2449
          AK294931.1 - 10, 11 2022
          AK295071.1 - 10, 11 2056
          AK304507.1 - 12, 13 2254
          AK304725.1 - 7, 8 763
          AK304774.1 - 8, 9 828
          AK314620.1 - 10, 11 2508
          AY039239.1 - 9, 10 818
          AY139007.1 - 2, 3 1153
          AY177411.1 - 8, 9 1219
          BC041391.1 - 12, 13 2618 5276767
          BX647696.1 - 10, 11 2453

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          cell cycle
          cell proliferation
          cerebral cortex development
          cell division
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0