Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC1 SIRNA Assays ›
Gene Symbol:
TACC1
Gene Name:
transforming acidic coiled-coil containing protein 1
Gene Aliases:
Ga55
Chromosome Location:
Chr.8: 38728186 - 38853028 on Build GRCh38
UniGene ID:
Hs.279245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS186180
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122824.1
NP_001116296.1
10
1467
NM_001146216.2
NP_001139688.1
12
2643
NM_006283.2
NP_006274.2
12
2697
XM_005273625.4
XP_005273682.1
14
2847
XM_005273628.1
XP_005273685.1
13
2354
XM_005273629.1
XP_005273686.1
14
2367
XM_005273630.1
XP_005273687.1
13
2375
XM_011544631.1
XP_011542933.1
13
2733
XM_011544632.2
XP_011542934.1
15
2908
XM_011544633.1
XP_011542935.1
13
2646
XM_011544634.1
XP_011542936.1
12
2610
XM_011544635.1
XP_011542937.1
13
2437
XM_011544636.2
XP_011542938.1
10
1093
XM_011544637.1
XP_011542939.1
10
1380
XM_011544638.2
XP_011542940.1
10
983
XM_017013766.1
XP_016869255.1
12
2318
XM_017013767.1
XP_016869256.1
13
2331
XM_017013768.1
XP_016869257.1
12
2231
XM_017013769.1
XP_016869258.1
13
2244
XM_017013770.1
XP_016869259.1
12
2267
XM_017013771.1
XP_016869260.1
12
2539
XM_017013772.1
XP_016869261.1
11
2231
XM_017013773.1
XP_016869262.1
11
2498
XM_017013774.1
XP_016869263.1
11
2183
XM_017013775.1
XP_016869264.1
12
2252
XM_017013776.1
XP_016869265.1
13
2244
XM_017013777.1
XP_016869266.1
11
2408
XM_017013778.1
XP_016869267.1
11
2107
GenBank mRNA
AB029026.1
-
11
1264
AF049910.1
-
12
2638
AK294931.1
-
12
2211
AK295071.1
-
12
2245
AK304507.1
-
14
2443
AK304725.1
-
9
952
AK304774.1
-
10
1017
AK314620.1
-
12
2697
AY039239.1
-
11
1007
AY139007.1
-
4
1342
AY177411.1
-
10
1408
BC041391.1
-
14
2807
5276767
BX647696.1
-
12
2642
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell cycle
cell proliferation
cerebral cortex development
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0