Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF168 SIRNA Assays ›
Gene Symbol:
RNF168
Gene Name:
ring finger protein 168
Gene Aliases:
hRNF168
Chromosome Location:
Chr.3: 196468783 - 196503768 on Build GRCh38
UniGene ID:
Hs.250648
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS186505
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF168
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152617.3
NP_689830.2
6
4126
GenBank mRNA
AK097068.1
-
1
1008
BC029433.1
-
1
123
BC047483.1
-
1
111
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
double-strand break repair via nonhomologous end joining
ubiquitin-dependent protein catabolic process
cellular response to DNA damage stimulus
response to ionizing radiation
protein ubiquitination
protein sumoylation
negative regulation of transcription elongation from RNA polymerase II promoter
histone H2A monoubiquitination
interstrand cross-link repair
histone H2A-K13 ubiquitination
histone H2A-K15 ubiquitination
isotype switching
positive regulation of DNA repair
protein K63-linked ubiquitination
histone H2A K63-linked ubiquitination
chromatin binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
nucleosome binding
histone binding
ubiquitin binding
K63-linked polyubiquitin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-kmb4w:80/100.66.76.53:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0