Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SHISA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145204.2 | NP_001138676.2 | 3 | 1174 | ||
XM_005255539.3 | XP_005255596.2 | 3 | 1175 | |||
XM_011522642.2 | XP_011520944.1 | 3 | 1175 | |||
GenBank mRNA | AK299610.1 | - | 4 | 604 |