Search
Search

MAP7D3| Gene Symbol: MAP7D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173516.1 | NP_001166987.1 | 10 | 1763 | ||
| NM_001173517.1 | NP_001166988.1 | 9 | 1770 | |||
| NM_024597.3 | NP_078873.2 | 10 | 1875 | |||
| XM_005262472.1 | XP_005262529.1 | 10 | 1727 | |||
| XM_017029843.1 | XP_016885332.1 | 10 | 1727 | |||
| GenBank mRNA | AK301478.1 | - | 10 | 1764 | ||
| AL832120.1 | - | 9 | 1562 | |||
| BC040518.1 | - | 10 | 1739 | |||
| BC064350.1 | - | 10 | 1555 | |||