Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CFAP69 SIRNA Assays ›
          Gene Symbol:
          CFAP69
          Gene Name:
          cilia and flagella associated protein 69
          Gene Aliases:
          C7orf63, FAP69
          Chromosome Location:
          Chr.7: 90245174 - 90320964 on Build GRCh38
          UniGene ID:
          Hs.521014
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS188222
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CFAP69
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039706.2 NP_001034795.2 11, 12 1414
          NM_001160138.1 NP_001153610.1 11, 12 1360
          XM_005250600.3 XP_005250657.1 11, 12 1393
          XM_011516577.2 XP_011514879.1 11, 12 1393
          XM_011516578.2 XP_011514880.1 11, 12 1393
          XM_011516579.1 XP_011514881.1 11, 12 1393
          XM_017012624.1 XP_016868113.1 11, 12 1339
          XM_017012625.1 XP_016868114.1 11, 12 1393
          XM_017012626.1 XP_016868115.1 11, 12 1393
          XM_017012627.1 XP_016868116.1 11, 12 1393
          XM_017012628.1 XP_016868117.1 11, 12 1393
          XM_017012629.1 XP_016868118.1 10, 11 1283
          XM_017012630.1 XP_016868119.1 11, 12 1393
          XM_017012631.1 XP_016868120.1 9, 10 941
          XM_017012632.1 XP_016868121.1 11, 12 1393
          XM_017012633.1 XP_016868122.1 9, 10 3970
          XM_017012634.1 XP_016868123.1 9, 10 1108
          XM_017012635.1 XP_016868124.1 7, 8 804
          XM_017012636.1 XP_016868125.1 6, 7 745
          XM_017012637.1 XP_016868126.1 7, 8 3910
          XM_017012638.1 XP_016868127.1 7, 8 751
          XM_017012639.1 XP_016868128.1 7, 8 786
          XM_017012640.1 XP_016868129.1 4, 5 390
          XM_017012641.1 XP_016868130.1 11, 12 1393
          XM_017012642.1 XP_016868131.1 11, 12 1393
          GenBank mRNA AK093991.1 - 9, 10 1196
          AK302636.1 - 11, 12 1319
          AK303034.1 - 11, 12 1393
          AL833446.1 - 4, 5 359
          BC036351.1 - 11, 12 1331
          BC132771.1 - 5, 6 636
          BC140742.1 - 11, 12 1300 9021610
          BC141834.1 - 11, 12 1360 40148850
          BC142655.1 - 11, 12 1360 40148060

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          structural protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0