Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG13 SIRNA Assays ›
Gene Symbol:
ALG13
Gene Name:
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases:
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location:
Chr.X: 111680741 - 111764164 on Build GRCh38
UniGene ID:
Hs.443061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS188236
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039210.4
NP_001034299.3
3
401
NM_001099922.2
NP_001093392.1
3
460
NM_001168385.2
NP_001161857.1
3
460
NM_001257230.1
NP_001244159.1
3
356
NM_001257231.1
NP_001244160.1
3
474
NM_001257234.1
NP_001244163.1
3
429
NM_001257235.2
NP_001244164.1
3
429
NM_001257237.1
NP_001244166.1
3
533
NM_001257239.2
NP_001244168.1
3
448
NM_001257240.2
NP_001244169.1
3
533
NM_001257241.2
NP_001244170.1
3
474
NM_001324290.1
NP_001311219.1
3
466
NM_001324291.1
NP_001311220.1
3
356
NM_001324292.1
NP_001311221.1
3
460
NM_001324293.1
NP_001311222.1
3
533
NM_001324294.1
NP_001311223.1
3
344
NM_018466.5
NP_060936.1
3
460
NR_033125.2
3
356
XM_006724693.2
XP_006724756.1
3
833
XM_006724697.3
XP_006724760.2
3
733
XM_006724698.3
XP_006724761.2
3
858
XM_011531028.2
XP_011529330.1
3
834
XM_011531030.1
XP_011529332.1
3
832
XM_011531031.1
XP_011529333.1
3
533
XM_011531032.1
XP_011529334.1
3
339
XM_011531033.2
XP_011529335.1
3
379
XM_017029846.1
XP_016885335.1
3
835
XM_017029847.1
XP_016885336.1
3
834
XM_017029848.1
XP_016885337.1
3
832
XM_017029849.1
XP_016885338.1
3
833
XM_017029850.1
XP_016885339.1
3
832
XM_017029851.1
XP_016885340.1
3
835
GenBank mRNA
AF220051.1
-
3
391
AK300394.1
-
3
298
AK302729.1
-
3
401
AK302890.1
-
3
377
AK304712.1
-
3
350
AK311608.1
-
3
340
AK312229.1
-
3
399
AK316306.1
-
3
402
AK316522.1
-
3
477
AL524204.3
-
3
279
BC005336.1
-
3
393
3996922
BE747995.1
-
3
389
BF239206.1
-
3
428
BF240663.1
-
3
246
BF699152.1
-
3
463
BG621042.1
-
2
243
BM759764.1
-
3
442
BQ007420.1
-
2
274
CB268871.1
-
3
407
DB292609.1
-
3
533
DC400909.1
-
3
344
DN992019.1
-
3
528
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
transferase
Gene Ontology Categories:
Function(s)
Process(es)
dolichol-linked oligosaccharide biosynthetic process
proteolysis
N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
thiol-dependent ubiquitinyl hydrolase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0