Search
Search

CCDC68| Gene Symbol: CCDC68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143829.1 | NP_001137301.1 | 9 | 883 | ||
| NM_025214.2 | NP_079490.1 | 9 | 986 | |||
| XM_006722552.3 | XP_006722615.1 | 9 | 935 | |||
| XM_011526198.2 | XP_011524500.1 | 9 | 1995 | |||
| XM_011526199.2 | XP_011524501.1 | 9 | 894 | |||
| XM_011526200.2 | XP_011524502.1 | 9 | 1047 | |||
| XM_011526201.2 | XP_011524503.1 | 9 | 1954 | |||
| XM_011526202.1 | XP_011524504.1 | 9 | 986 | |||
| XM_011526203.2 | XP_011524505.1 | 8 | 821 | |||
| XM_017026011.1 | XP_016881500.1 | 8 | 987 | |||
| GenBank mRNA | AF273051.1 | - | 9 | 884 | ||
| AK058097.1 | - | 9 | 883 | |||
| AK313793.1 | - | 7 | 729 | |||
| BC029508.1 | - | 9 | 940 | 5297655 | ||