Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OBSCN SIRNA Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
UniGene ID:
Hs.650039
Species:
Human
siRNA ID
HSS188886
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OBSCN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098623.2
NP_001092093.2
55
15370
NM_001271223.2
NP_001258152.2
66
18241
NM_052843.3
NP_443075.3
55
15370
XM_005273287.4
XP_005273344.1
67
18621
XM_005273291.4
XP_005273348.1
67
18585
XM_005273298.4
XP_005273355.1
66
18312
XM_005273307.4
XP_005273364.1
62
17499
XM_006711822.3
XP_006711885.1
65
18045
XM_006711823.3
XP_006711886.1
65
18045
XM_006711827.3
XP_006711890.1
59
16460
XM_006711829.3
XP_006711892.1
58
16184
XM_011544297.2
XP_011542599.1
59
16461
XM_011544299.2
XP_011542601.1
58
16185
XM_017002443.1
XP_016857932.1
67
18666
XM_017002444.1
XP_016857933.1
67
18629
XM_017002445.1
XP_016857934.1
67
18666
XM_017002446.1
XP_016857935.1
67
18666
XM_017002447.1
XP_016857936.1
67
18663
XM_017002448.1
XP_016857937.1
67
18516
XM_017002449.1
XP_016857938.1
66
18403
XM_017002450.1
XP_016857939.1
66
18403
XM_017002451.1
XP_016857940.1
66
18403
XM_017002452.1
XP_016857941.1
66
18403
XM_017002453.1
XP_016857942.1
66
18390
XM_017002454.1
XP_016857943.1
66
18390
XM_017002455.1
XP_016857944.1
66
18390
XM_017002456.1
XP_016857945.1
66
18390
XM_017002457.1
XP_016857946.1
66
18390
XM_017002458.1
XP_016857947.1
65
18139
XM_017002459.1
XP_016857948.1
65
18139
XM_017002460.1
XP_016857949.1
65
18139
XM_017002461.1
XP_016857950.1
65
18126
XM_017002462.1
XP_016857951.1
65
18114
XM_017002463.1
XP_016857952.1
65
18093
XM_017002464.1
XP_016857953.1
65
18042
XM_017002465.1
XP_016857954.1
64
17875
XM_017002466.1
XP_016857955.1
64
17875
XM_017002467.1
XP_016857956.1
64
17875
XM_017002468.1
XP_016857957.1
63
17611
XM_017002469.1
XP_016857958.1
61
17083
XM_017002470.1
XP_016857959.1
56
15764
XM_017002471.1
XP_016857960.1
67
18665
XM_017002472.1
XP_016857961.1
67
18665
XM_017002473.1
XP_016857962.1
67
18665
GenBank mRNA
AJ002535.1
-
56
15367
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
multicellular organism development
regulation of Rho protein signal transduction
protein localization to M-band
positive regulation of apoptotic process
positive regulation of GTPase activity
sarcomere organization
regulation of small GTPase mediated signal transduction
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
ankyrin binding
titin binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-jgtjs:80/10.239.162.121:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0