Search
Search

SLC41A2| Gene Symbol: SLC41A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032148.3 | NP_115524.3 | 7 | 1391 | ||
| XM_005269176.1 | XP_005269233.1 | 8 | 1597 | |||
| XM_005269178.1 | XP_005269235.1 | 8 | 1492 | |||
| XM_005269179.1 | XP_005269236.1 | 8 | 1429 | |||
| XM_006719630.2 | XP_006719693.1 | 9 | 1752 | |||
| XM_011538807.2 | XP_011537109.1 | 9 | 1693 | |||
| XM_011538808.1 | XP_011537110.1 | 9 | 1702 | |||
| XM_011538809.1 | XP_011537111.1 | 9 | 1649 | |||
| XM_011538810.2 | XP_011537112.1 | 8 | 1519 | |||
| XM_011538811.2 | XP_011537113.1 | 8 | 1975 | |||
| XM_011538813.2 | XP_011537115.1 | 6 | 826 | |||
| XM_017020013.1 | XP_016875502.1 | 8 | 1514 | |||
| XM_017020014.1 | XP_016875503.1 | 9 | 1752 | |||
| XM_017020016.1 | XP_016875505.1 | 8 | 874 | |||
| XM_017020017.1 | XP_016875506.1 | 6 | 2142 | |||
| GenBank mRNA | AK027838.1 | - | 7 | 1316 | ||
| AL136828.1 | - | 7 | 1083 | |||
| BC036734.1 | - | 7 | 1363 | 5538264 | ||
| BC106871.1 | - | 7 | 1308 | 40006660 | ||
| BC106872.1 | - | 7 | 1308 | 40006662 | ||