Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281302.1 | NP_001268231.1 | 7 | 946 | ||
NM_032380.4 | NP_115756.2 | 6 | 828 | |||
NM_170681.2 | NP_733781.1 | 6 | 828 | |||
NM_170691.2 | NP_733792.1 | 6 | 828 | |||
NR_104006.1 | 6 | 828 | ||||
XM_011543691.2 | XP_011541993.1 | 6 | 611 | |||
XM_017009986.1 | XP_016865475.1 | 6 | 722 | |||
GenBank mRNA | AF111808.1 | - | 6 | 578 | ||
AF367997.1 | - | 6 | 674 | |||
AK025314.1 | - | 6 | 605 | |||
AK308422.1 | - | 6 | 575 | |||
AK315705.1 | - | 6 | 607 | |||
AY453587.1 | - | 6 | 572 | |||
BC015712.1 | - | 7 | 680 | 3922036 | ||
BC030612.1 | - | 6 | 577 | 4811939 |