Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM2 SIRNA Assays ›
Gene Symbol:
ABLIM2
Gene Name:
actin binding LIM protein family member 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 7965310 - 8158832 on Build GRCh38
UniGene ID:
Hs.233404
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS189073
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130083.1
NP_001123555.1
2
282
NM_001130084.1
NP_001123556.1
2
282
NM_001130085.1
NP_001123557.1
2
282
NM_001130086.1
NP_001123558.1
2
282
NM_001130087.1
NP_001123559.1
2
282
NM_001130088.1
NP_001123560.1
2
282
NM_032432.4
NP_115808.3
2
282
XM_005248014.4
XP_005248071.1
2
253
XM_005248017.4
XP_005248074.1
2
253
XM_005248018.4
XP_005248075.1
2
252
XM_005248019.4
XP_005248076.1
2
252
XM_005248020.4
XP_005248077.1
2
252
XM_005248021.4
XP_005248078.1
2
251
XM_005248022.4
XP_005248079.1
2
252
XM_005248023.4
XP_005248080.1
2
252
XM_005248024.4
XP_005248081.1
2
251
XM_005248027.4
XP_005248084.1
2
251
XM_005248028.4
XP_005248085.1
2
248
XM_005248029.4
XP_005248086.1
2
250
XM_005248030.4
XP_005248087.1
2
250
XM_005248031.4
XP_005248088.1
2
252
XM_006713923.3
XP_006713986.1
2
253
XM_006713924.3
XP_006713987.1
2
252
XM_006713925.3
XP_006713988.1
2
253
XM_006713927.3
XP_006713990.1
2
251
XM_011513586.2
XP_011511888.1
2
257
XM_017008706.1
XP_016864195.1
2
254
XM_017008707.1
XP_016864196.1
2
263
XM_017008708.1
XP_016864197.1
2
252
XM_017008709.1
XP_016864198.1
2
253
XM_017008710.1
XP_016864199.1
2
252
XM_017008711.1
XP_016864200.1
2
252
XM_017008712.1
XP_016864201.1
2
252
XM_017008713.1
XP_016864202.1
2
251
XM_017008714.1
XP_016864203.1
2
263
XM_017008715.1
XP_016864204.1
2
252
XM_017008716.1
XP_016864205.1
2
251
XM_017008717.1
XP_016864206.1
2
251
XM_017008718.1
XP_016864207.1
2
263
XM_017008719.1
XP_016864208.1
2
251
XM_017008720.1
XP_016864209.1
2
250
XM_017008721.1
XP_016864210.1
2
250
XM_017008722.1
XP_016864211.1
2
251
XM_017008723.1
XP_016864212.1
2
263
XM_017008724.1
XP_016864213.1
2
263
XM_017008725.1
XP_016864214.1
2
263
GenBank mRNA
AB058711.1
-
1
41
AJ748600.1
-
2
204
AJ748601.1
-
2
204
AK094754.1
-
2
261
AK301060.1
-
2
245
BC067214.1
-
2
294
BC122567.1
-
2
233
40085570
DQ413176.1
-
2
159
DQ413177.1
-
1
139
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
actin cytoskeleton organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qwrcb:80/100.66.75.127:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0