Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN5 SIRNA Assays ›
          Gene Symbol:
          PTPN5
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 5
          Gene Aliases:
          PTPSTEP, STEP, STEP61
          Chromosome Location:
          Chr.11: 18727928 - 18792721 on Build GRCh38
          UniGene ID:
          Hs.79092
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS189231
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPN5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039970.1 NP_001035059.1 10 1384
          NM_001278236.1 NP_001265165.1 10 2218
          NM_001278238.1 NP_001265167.1 9 1347
          NM_001278239.1 NP_001265168.1 9 1251
          NM_006906.1 NP_008837.1 10 1480
          NM_032781.3 NP_116170.3 10 1435
          XM_011520411.2 XP_011518713.1 10 1419
          XM_017018434.1 XP_016873923.1 10 1419
          XM_017018435.1 XP_016873924.1 10 1389
          XM_017018436.1 XP_016873925.1 9 1325
          XM_017018437.1 XP_016873926.1 10 1343
          XM_017018438.1 XP_016873927.1 8 1104
          XM_017018439.1 XP_016873928.1 9 1230
          XM_017018440.1 XP_016873929.1 10 1419
          XM_017018441.1 XP_016873930.1 10 1419
          GenBank mRNA AK090923.1 - 10 1344
          AK127312.1 - 10 2218
          AK294374.1 - 9 1087
          AK294673.1 - 9 1048
          AK295604.1 - 9 1208
          AK316270.1 - 9 1132
          AL832541.1 - 10 1480
          BC039897.1 - 9 1143
          BC064807.1 - 10 1244 5726473

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          response to amphetamine
          positive regulation of receptor internalization
          protein dephosphorylation
          positive regulation of neuron projection development
          positive regulation of protein import into nucleus, translocation
          peptidyl-tyrosine dephosphorylation
          exploration behavior
          response to immobilization stress
          response to drug
          negative regulation of MAP kinase activity
          positive regulation of long-term synaptic potentiation
          positive regulation of neuron death
          response to acetylsalicylate
          positive regulation of protein targeting to mitochondrion
          positive regulation of response to drug
          phosphotyrosine binding
          protein tyrosine phosphatase activity
          protein binding
          glutamate receptor binding
          mitogen-activated protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0