This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
GEMIN2
- Gene Name:
-
gem nuclear organelle associated protein 2
- Gene Aliases:
-
SIP1, SIP1-delta
- Chromosome Location:
-
Chr.14: 39114261 - 39136973 on Build GRCh38
- UniGene ID:
-
Hs.652307
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
GEMIN2 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_001009182.1 |
NP_001009182.1 |
3, 4
|
372
|
|
| NM_001009183.1 |
NP_001009183.1 |
3, 4
|
372
|
|
| NM_003616.2 |
NP_003607.1 |
3, 4
|
372
|
|
| XM_017021709.1 |
XP_016877198.1 |
3, 4
|
395
|
|
| GenBank mRNA |
AB037701.1 |
- |
3, 4
|
350
|
|
| AB037702.1 |
- |
3, 4
|
350
|
|
| AB037703.1 |
- |
3, 4
|
270
|
|
| AF027150.1 |
- |
3, 4
|
433
|
|
| AK313947.1 |
- |
3, 4
|
360
|
|
| BC028095.1 |
- |
3, 4
|
371
|
|
| BC104968.1 |
- |
3, 4
|
371
|
8143971 |
| CN484054.1 |
- |
3, 4
|
372
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System