Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145343.1 | NP_001138815.1 | 5 | 615 | ||
NM_001145344.1 | NP_001138816.1 | 5 | 612 | |||
NM_001145345.1 | NP_001138817.1 | 5 | 624 | |||
NM_001300970.1 | NP_001287899.1 | 4 | 838 | |||
NM_032838.4 | NP_116227.1 | 5 | 559 | |||
XM_005259354.2 | XP_005259411.1 | 4 | 857 | |||
XM_005259356.2 | XP_005259413.1 | 3 | 742 | |||
XM_006723447.2 | XP_006723510.1 | 5 | 968 | |||
XM_011527428.1 | XP_011525730.1 | 3 | 730 | |||
XM_017027399.1 | XP_016882888.1 | 4 | 826 | |||
XM_017027400.1 | XP_016882889.1 | 2 | 387 | |||
GenBank mRNA | AK027685.1 | - | 5 | 559 | ||
AK074497.1 | - | 5 | 569 | |||
AK299393.1 | - | 3 | 389 | |||
AK316262.1 | - | 3 | 379 | |||
BC007064.2 | - | 4 | 536 | 3961390 | ||
BC104886.1 | - | 4 | 531 | 8143889 | ||
BC104888.1 | - | 4 | 531 | 8143891 | ||
BC143658.1 | - | 4 | 534 | 9052172 | ||
BX649070.1 | - | 3 | 690 |