Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039374.4 | NP_001034463.4 | 11 | 1289 | ||
GenBank mRNA | AK090683.1 | - | 6 | 1129 | ||
AK092639.1 | - | 7 | 1812 | |||
AK094220.1 | - | 1 | 975 | |||
BC064582.1 | - | 11 | 1275 | |||
BC137317.1 | - | 11 | 1264 | 9021314 | ||
BC137318.1 | - | 11 | 1264 | 9021315 |