Search
Search

PSMG1| Gene Symbol: PSMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261824.1 | NP_001248753.1 | 5 | 723 | ||
| NM_001320795.1 | NP_001307724.1 | 5 | 926 | |||
| NM_003720.3 | NP_003711.1 | 5 | 738 | |||
| NM_203433.2 | NP_982257.1 | 4 | 675 | |||
| NR_049728.1 | 4 | 631 | ||||
| XM_017028488.1 | XP_016883977.1 | 4 | 589 | |||
| GenBank mRNA | AB451468.1 | - | 4 | 546 | ||
| AF417108.1 | - | 4 | 502 | |||
| AJ006291.1 | - | 5 | 672 | |||
| AK001455.1 | - | 5 | 700 | |||
| AK308614.1 | - | 4 | 609 | |||
| AK315406.1 | - | 5 | 640 | |||
| AW594615.1 | - | 3 | 400 | |||
| AY463963.1 | - | 4 | 546 | |||
| BC003619.2 | - | 5 | 655 | 2961575 | ||
| BC010424.1 | - | 5 | 647 | 3343732 | ||
| BC011755.2 | - | 5 | 655 | 3531528 | ||
| BC012809.2 | - | 5 | 655 | 2961575 | ||
| BM013494.1 | - | 5 | 608 | |||
| BM460414.1 | - | 5 | 631 | |||
| BM462074.1 | - | 5 | 655 | |||
| BQ008489.1 | - | 3 | 403 | |||
| CR541821.1 | - | 5 | 609 | |||
| CR541852.1 | - | 5 | 609 | |||