Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KYAT1 SIRNA Assays ›
Gene Symbol:
KYAT1
Gene Name:
kynurenine aminotransferase 1
Gene Aliases:
CCBL1, GTK, KAT1, KATI
Chromosome Location:
Chr.9: 128832942 - 128882551 on Build GRCh38
UniGene ID:
Hs.495250
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS189612
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KYAT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122671.1
NP_001116143.1
12
1334
NM_001122672.1
NP_001116144.1
11
1190
NM_001287390.1
NP_001274319.1
14
1731
NM_004059.4
NP_004050.3
12
1340
NR_109829.1
12
1372
XM_006717311.3
XP_006717374.1
13
1526
XM_011519165.2
XP_011517467.1
14
1702
XM_011519167.2
XP_011517469.2
12
1306
XM_011519168.1
XP_011517470.1
11
1165
XM_011519169.1
XP_011517471.1
12
1434
XM_011519170.2
XP_011517472.1
12
1802
XM_017015260.1
XP_016870749.1
13
1486
XM_017015261.1
XP_016870750.1
14
1708
XM_017015262.1
XP_016870751.1
14
2197
XM_017015263.1
XP_016870752.1
14
1612
XM_017015264.1
XP_016870753.1
13
1519
XM_017015265.1
XP_016870754.1
12
1896
XM_017015266.1
XP_016870755.1
13
1527
XM_017015267.1
XP_016870756.1
12
1348
XM_017015268.1
XP_016870757.1
13
1510
GenBank mRNA
AK094505.1
-
12
1915
AK291178.1
-
12
1328
AK297995.1
-
14
1587
AK314427.1
-
12
1210
BC021262.2
-
12
1333
4137942
BC033685.1
-
11
1044
5193675
BU622726.1
-
2
599
X82224.1
-
11
1155
Back To Top
More Information
Panther Classification:
Molecular Function -
transaminase
Gene Ontology Categories:
Function(s)
Process(es)
L-phenylalanine catabolic process
tryptophan catabolic process
cellular modified amino acid metabolic process
cellular amino acid biosynthetic process
kynurenine metabolic process
L-kynurenine catabolic process
protein binding
transaminase activity
kynurenine-oxoglutarate transaminase activity
pyridoxal phosphate binding
protein homodimerization activity
L-phenylalanine:pyruvate aminotransferase activity
glutamine-phenylpyruvate transaminase activity
cysteine-S-conjugate beta-lyase activity
L-glutamine:pyruvate aminotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8lnnk:80/100.66.76.235:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0