Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PLCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033123.3 | NP_149114.2 | 7 | 1088 | ||
NR_073075.1 | 7 | 1091 | ||||
XM_005253519.2 | XP_005253576.1 | 7 | 1081 | |||
XM_011520883.1 | XP_011519185.1 | 4 | 494 | |||
XM_017020176.1 | XP_016875665.1 | 7 | 1081 | |||
XM_017020177.1 | XP_016875666.1 | 7 | 1081 | |||
XM_017020179.1 | XP_016875668.1 | 7 | 1076 | |||
XM_017020180.1 | XP_016875669.1 | 7 | 970 | |||
XM_017020181.1 | XP_016875670.1 | 7 | 1035 | |||
XM_017020182.1 | XP_016875671.1 | 3 | 734 | |||
XM_017020183.1 | XP_016875672.1 | 4 | 498 | |||
XM_017020184.1 | XP_016875673.1 | 4 | 426 | |||
XM_017020185.1 | XP_016875674.1 | 3 | 311 | |||
XM_017020186.1 | XP_016875675.1 | 4 | 456 | |||
GenBank mRNA | AF532185.1 | - | 7 | 1013 | ||
AK097555.1 | - | 3 | 436 | |||
AK097725.1 | - | 4 | 431 | |||
AK292279.1 | - | 7 | 1086 | |||
AK314516.1 | - | 7 | 1045 | |||
AY035866.1 | - | 7 | 1050 | |||
BC125067.1 | - | 3 | 341 | 40117397 |