Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: SYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197129.1 | NP_001184058.1 | 5 | 566 | ||
NM_033542.3 | NP_291020.1 | 4 | 542 | |||
GenBank mRNA | AK075264.1 | - | 4 | 505 | ||
AK309429.1 | - | 4 | 484 | |||
AL591714.1 | - | 4 | 437 | |||
BC048286.1 | - | 4 | 472 | 5212523 | ||
BI820996.1 | - | 5 | 566 | |||
DA526637.1 | - | 4 | 542 |