Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP7 SIRNA Assays ›
Gene Symbol:
MAP7
Gene Name:
microtubule associated protein 7
Gene Aliases:
E-MAP-115, EMAP115
Chromosome Location:
Chr.6: 136342281 - 136551015 on Build GRCh38
UniGene ID:
Hs.486548
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS189817
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198608.1
NP_001185537.1
3, 4
556
NM_001198609.1
NP_001185538.1
3, 4
556
NM_001198611.1
NP_001185540.1
3, 4
556
NM_001198614.1
NP_001185543.1
4, 5
559
NM_001198615.1
NP_001185544.1
3, 4
289
NM_001198616.1
NP_001185545.1
3, 4
652
NM_001198618.1
NP_001185547.1
3, 4
419
NM_001198619.1
NP_001185548.1
3, 4
292
NM_003980.4
NP_003971.1
3, 4
652
XM_006715598.2
XP_006715661.1
4, 5
1077
XM_006715599.2
XP_006715662.1
4, 5
846
XM_006715600.2
XP_006715663.1
4, 5
846
XM_006715601.3
XP_006715664.1
3, 4
848
XM_011536243.2
XP_011534545.1
3, 4
1447
XM_011536244.2
XP_011534546.1
3, 4
1467
XM_011536245.1
XP_011534547.1
3, 4
346
XM_011536246.2
XP_011534548.1
3, 4
1448
XM_017011471.1
XP_016866960.1
4, 5
1077
GenBank mRNA
AJ242502.1
-
3, 4
241
AK294461.1
-
3, 4
326
AK295749.1
-
3, 4
284
AK296483.1
-
3, 4
419
AK296556.1
-
3, 4
418
AK299355.1
-
3, 4
289
AK307873.1
-
3, 4
395
AK311227.1
-
3, 4
571
AK311234.1
-
4, 5
436
AK311236.1
-
4, 5
394
AK311270.1
-
4, 5
559
AK312789.1
-
3, 4
252
AK316062.1
-
3, 4
292
AK316529.1
-
4, 5
425
BC025777.1
-
3, 4
248
5222987
CD557798.1
-
3, 4
492
DA329576.1
-
3, 4
463
DC325462.1
-
3, 4
506
X73882.1
-
3, 4
341
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
response to osmotic stress
establishment or maintenance of cell polarity
protein localization to plasma membrane
receptor binding
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-blthd:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline