Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACVR1 SIRNA Assays ›
          Gene Symbol:
          ACVR1
          Gene Name:
          activin A receptor type 1
          Gene Aliases:
          ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI
          Chromosome Location:
          Chr.2: 157736446 - 157875880 on Build GRCh38
          UniGene ID:
          Hs.470316
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS189883
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACVR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105.4 NP_001096.1 8 1339
          NM_001111067.2 NP_001104537.1 8 1158
          XM_005246939.3 XP_005246996.1 8 1210
          XM_005246940.3 XP_005246997.1 8 1178
          XM_006712825.3 XP_006712888.1 8 1291
          XM_011512106.2 XP_011510408.1 9 1258
          XM_011512107.2 XP_011510409.1 8 1230
          XM_011512108.2 XP_011510410.1 8 2637
          GenBank mRNA AK313630.1 - 8 1134
          BC033867.1 - 8 1339 5209711
          BC036748.1 - 8 1339
          KU177885.1 - 6 909
          KU177886.1 - 6 909
          L02911.1 - 9 1249
          Z22534.1 - 7 1012

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          patterning of blood vessels
          in utero embryonic development
          gastrulation with mouth forming second
          mesoderm formation
          neural crest cell migration
          acute inflammatory response
          embryonic heart tube morphogenesis
          outflow tract septum morphogenesis
          atrioventricular valve morphogenesis
          mitral valve morphogenesis
          endocardial cushion morphogenesis
          endocardial cushion fusion
          atrial septum primum morphogenesis
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          germ cell development
          determination of left/right symmetry
          negative regulation of signal transduction
          positive regulation of pathway-restricted SMAD protein phosphorylation
          peptidyl-threonine phosphorylation
          signal transduction by protein phosphorylation
          regulation of ossification
          positive regulation of cell migration
          positive regulation of bone mineralization
          BMP signaling pathway
          activin receptor signaling pathway
          negative regulation of activin receptor signaling pathway
          positive regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          smooth muscle cell differentiation
          pharyngeal system development
          pathway-restricted SMAD protein phosphorylation
          ventricular septum morphogenesis
          cardiac muscle cell fate commitment
          BMP signaling pathway involved in heart development
          endocardial cushion cell fate commitment
          cellular response to glucocorticoid stimulus
          cellular response to BMP stimulus
          positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          positive regulation of determination of dorsal identity
          negative regulation of extrinsic apoptotic signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          transmembrane receptor protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta receptor activity, type I
          protein binding
          ATP binding
          activin receptor activity, type I
          peptide hormone binding
          protein homodimerization activity
          SMAD binding
          metal ion binding
          activin binding
          transforming growth factor beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0