Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NREP SIRNA Assays ›
Gene Symbol:
NREP
Gene Name:
neuronal regeneration related protein
Gene Aliases:
C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17
Chromosome Location:
Chr.5: 111729303 - 111976931 on Build GRCh38
UniGene ID:
Hs.36053
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS190184
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NREP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142474.1
NP_001135946.1
4
401
NM_001142475.1
NP_001135947.1
4
431
NM_001142476.1
NP_001135948.1
5
483
NM_001142477.1
NP_001135949.1
5
374
NM_001142478.1
NP_001135950.1
5
350
NM_001142479.1
NP_001135951.1
5
387
NM_001142480.1
NP_001135952.1
5
381
NM_001142481.1
NP_001135953.1
5
290
NM_001142482.1
NP_001135954.1
5
297
NM_001142483.1
NP_001135955.1
3
586
NM_004772.2
NP_004763.1
4
278
XM_006714732.1
XP_006714795.1
3
198
XM_006714733.2
XP_006714796.1
4
179
GenBank mRNA
AK298779.1
-
4
326
AK309667.1
-
3
108
AK315617.1
-
4
252
AL547663.3
-
5
378
BC011050.1
-
5
311
4340005
BC019068.2
-
4
214
4893075
BC072013.1
-
4
237
5922876
BC072443.1
-
4
242
6162335
DA027062.1
-
4
401
DA078027.1
-
3
311
DA490948.1
-
5
483
DA520221.1
-
5
309
DA526263.1
-
5
471
DA683910.1
-
5
297
DA694580.1
-
4
296
DA704824.1
-
5
387
DA744980.1
-
5
234
DA752769.1
-
5
290
DA908653.1
-
4
278
DA939543.1
-
5
374
DC365764.1
-
4
431
U30521.1
-
5
309
U36189.1
-
3
122
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transforming growth factor beta receptor signaling pathway
axon regeneration
regulation of neuron differentiation
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0