Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: STOML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256672.1 | NP_001243601.1 | 7 | 1157 | ||
NM_001256673.1 | NP_001243602.1 | 6 | 1010 | |||
NM_001256674.1 | NP_001243603.1 | 6 | 1007 | |||
NM_001256675.1 | NP_001243604.1 | 6 | 947 | |||
NM_001256676.1 | NP_001243605.1 | 7 | 971 | |||
NM_001256677.1 | NP_001243606.1 | 8 | 1288 | |||
NM_001324226.1 | NP_001311155.1 | 6 | 1276 | |||
NM_001324227.1 | NP_001311156.1 | 7 | 974 | |||
NM_001324228.1 | NP_001311157.1 | 7 | 1038 | |||
NM_001324229.1 | NP_001311158.1 | 6 | 1465 | |||
NM_001324230.1 | NP_001311159.1 | 8 | 1291 | |||
NM_004809.4 | NP_004800.2 | 7 | 1160 | |||
XM_005254792.4 | XP_005254849.1 | 7 | 2064 | |||
XM_006720773.3 | XP_006720836.1 | 7 | 2062 | |||
GenBank mRNA | AF074953.1 | - | 7 | 1022 | ||
AF111800.1 | - | 6 | 959 | |||
AF156557.1 | - | 7 | 1168 | |||
AK075244.1 | - | 7 | 1056 | |||
AK093689.1 | - | 3 | 1425 | |||
AK222665.1 | - | 7 | 1040 | |||
AK300799.1 | - | 8 | 1172 | |||
AK302051.1 | - | 6 | 965 | |||
AL109664.1 | - | 6 | 837 | |||
AL109665.1 | - | 7 | 1011 | |||
AL533460.3 | - | 7 | 948 | |||
BC034379.1 | - | 7 | 1106 | 5185908 | ||
BC065249.1 | - | 7 | 1106 | 6059393 | ||
CR542089.1 | - | 7 | 1035 | |||
DQ064605.1 | - | 6 | 822 | |||
Y16522.1 | - | 7 | 1067 |