Search
Search

AU040320| Gene Symbol: AU040320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035525.1 | NP_001030602.1 | 12 | 2019 | ||
| NM_001035526.1 | NP_001030603.1 | 12 | 2137 | |||
| NM_133886.2 | NP_598647.1 | 12 | 2137 | |||
| GenBank mRNA | AK031342.1 | - | 12 | 2081 | ||
| AK043006.1 | - | 12 | 2087 | |||
| AK049570.1 | - | 12 | 2020 | |||
| AK084668.1 | - | 12 | 1982 | |||
| AK147569.1 | - | 4668 | ||||
| AK161493.1 | - | 2 | 200 | |||
| AK170261.1 | - | 12 | 2094 | |||
| AK198153.1 | - | 2 | 135 | |||
| AK211858.1 | - | 2 | 220 | |||
| BC022154.1 | - | 3 | 549 | |||
| BC028869.1 | - | 12 | 2107 | 4235813 | ||
| CA540237.1 | - | 3 | 443 | |||