Search
Search

Papd4| Gene Symbol: Papd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133905.2 | NP_598666.1 | 6 | 833 | ||
| GenBank mRNA | AK033141.1 | - | 3 | 170 | ||
| AK146086.1 | - | 6 | 817 | |||
| AM419011.1 | - | 5 | 606 | |||
| BC016629.1 | - | 6 | 833 | 3595512 | ||