Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADIPOQ SIRNA Assays ›
          Gene Symbol:
          Adipoq
          Gene Name:
          adiponectin, C1Q and collagen domain containing
          Gene Aliases:
          30kDa, APN, Acdc, Acrp30, Ad, GBP28, adipo, apM1
          Chromosome Location:
          Chr.16: 23146536 - 23157968 on Build GRCm38
          UniGene ID:
          Mm.3969
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS201612
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Adipoq
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009605.4 NP_033735.3 2 228
          GenBank mRNA AK003138.1 - 2 146
          AK041214.1 - 2 178
          AK134112.1 - 2 228
          AX767955.1 - 1 130
          AX767957.1 - 2 159
          AY749429.1 - 1 114
          AY754346.1 - 1 114
          BC028770.1 - 2 143 1329799
          U37222.1 - 2 159
          U49915.1 - 1 130

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          glucose metabolic process
          fatty acid beta-oxidation
          response to glucose
          positive regulation of signal transduction
          negative regulation of platelet-derived growth factor receptor signaling pathway
          positive regulation of protein kinase A signaling
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of cholesterol efflux
          regulation of glucose metabolic process
          negative regulation of smooth muscle cell migration
          fatty acid oxidation
          negative regulation of cell migration
          negative regulation of granulocyte differentiation
          negative regulation of protein autophosphorylation
          positive regulation of cellular protein metabolic process
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-8 production
          cellular response to insulin stimulus
          positive regulation of myeloid cell apoptotic process
          adiponectin-activated signaling pathway
          negative regulation of heterotypic cell-cell adhesion
          low-density lipoprotein particle clearance
          response to tumor necrosis factor
          cellular response to drug
          glucose homeostasis
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of MAP kinase activity
          negative regulation of fat cell differentiation
          negative regulation of macrophage differentiation
          negative regulation of low-density lipoprotein particle receptor biosynthetic process
          negative regulation of gluconeogenesis
          negative regulation of blood pressure
          positive regulation of blood pressure
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of fatty acid metabolic process
          positive regulation of glucose import
          negative regulation of hormone secretion
          negative regulation of smooth muscle cell proliferation
          negative regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of phagocytosis
          negative regulation of synaptic transmission
          brown fat cell differentiation
          protein homooligomerization
          membrane depolarization
          membrane hyperpolarization
          protein heterotrimerization
          negative regulation of ERK1 and ERK2 cascade
          detection of oxidative stress
          cellular response to cAMP
          positive regulation of monocyte chemotactic protein-1 production
          cellular response to epinephrine stimulus
          protein localization to plasma membrane
          negative regulation of intracellular protein transport
          negative regulation of receptor binding
          negative regulation of DNA biosynthetic process
          positive regulation of glycogen (starch) synthase activity
          positive regulation of metanephric glomerular visceral epithelial cell development
          positive regulation of cAMP-dependent protein kinase activity
          positive regulation of renal albumin absorption
          negative regulation of platelet-derived growth factor receptor-alpha signaling pathway
          negative regulation of metanephric mesenchymal cell migration
          receptor binding
          hormone activity
          protein binding
          sialic acid binding
          identical protein binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0