Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADRB2 SIRNA Assays ›
          Gene Symbol:
          Adrb2
          Gene Name:
          adrenergic receptor, beta 2
          Gene Aliases:
          Adrb-2, Badm, Gpcr7
          Chromosome Location:
          Chr.18: 62177713 - 62179981 on Build GRCm38
          UniGene ID:
          Mm.5598
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS201799
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Adrb2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007420.3 NP_031446.2 1 872
          GenBank mRNA AK080241.1 - 1 870
          AK080276.1 - 1 872
          BC030346.1 - 1 308
          BC032883.1 - 1 850 1349283

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of systemic arterial blood pressure by norepinephrine-epinephrine
          diet induced thermogenesis
          vasodilation by norepinephrine-epinephrine involved in regulation of systemic arterial blood pressure
          regulation of sodium ion transport
          desensitization of G-protein coupled receptor protein signaling pathway by arrestin
          diaphragm contraction
          positive regulation of the force of heart contraction by epinephrine
          receptor-mediated endocytosis
          regulation of smooth muscle contraction
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          cell-cell signaling
          positive regulation of cell proliferation
          associative learning
          response to cold
          positive regulation of sodium ion transport
          negative regulation of angiogenesis
          negative regulation of ossification
          positive regulation of bone mineralization
          positive regulation of protein ubiquitination
          heat generation
          positive regulation of ATPase activity
          synaptic transmission, glutamatergic
          negative regulation of urine volume
          negative regulation of multicellular organism growth
          wound healing
          regulation of vasodilation
          positive regulation of apoptotic process
          positive regulation of potassium ion transport
          positive regulation of MAPK cascade
          bone resorption
          positive regulation of heart contraction
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of smooth muscle contraction
          positive regulation of skeletal muscle tissue growth
          negative regulation of inflammatory response
          brown fat cell differentiation
          regulation of calcium ion transport
          regulation of sensory perception of pain
          excitatory postsynaptic potential
          cellular response to hypoxia
          adrenergic receptor signaling pathway
          adenylate cyclase-activating adrenergic receptor signaling pathway
          negative regulation of platelet aggregation
          mitophagy in response to mitochondrial depolarization
          G-protein alpha-subunit binding
          signal transducer activity
          G-protein coupled receptor activity
          adrenergic receptor activity
          beta2-adrenergic receptor activity
          protein binding
          drug binding
          adenylate cyclase binding
          potassium channel regulator activity
          enzyme binding
          B2 bradykinin receptor binding
          protein complex binding
          dopamine binding
          ionotropic glutamate receptor binding
          protein homodimerization activity
          epinephrine binding
          norepinephrine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0