Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP2B2 SIRNA Assays ›
          Gene Symbol:
          Atp2b2
          Gene Name:
          ATPase, Ca++ transporting, plasma membrane 2
          Gene Aliases:
          D6Abb2e, Gena300, PMCA2, Tmy, dfw, jog, wms, wri
          Chromosome Location:
          Chr.6: 113743842 - 114042613 on Build GRCm38
          UniGene ID:
          Mm.321755
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS202252
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Atp2b2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001036684.2 NP_001031761.1 8 1609
          NM_009723.5 NP_033853.1 10 1798
          GenBank mRNA AB030737.1 - 7 1170
          AB030738.1 - 7 1170
          AF053471.1 - 9 1627
          AK054112.1 - 12 1895
          AK147240.1 - 10 1684
          AK147357.1 - 8 1538
          AK147369.1 - 10 1734
          AK147399.1 - 8 1773
          AK147501.1 - 10 1745
          BC075643.1 - 8 1609 30621322
          GU734816.2 - 10 1305

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          transport
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          organelle organization
          lactation
          sensory perception of sound
          locomotory behavior
          regulation of cell size
          cerebellum development
          cerebellar Purkinje cell layer morphogenesis
          cerebellar Purkinje cell differentiation
          cerebellar granule cell differentiation
          neuron differentiation
          locomotion
          serotonin metabolic process
          inner ear morphogenesis
          otolith mineralization
          cGMP metabolic process
          regulation of synaptic plasticity
          inner ear development
          synapse organization
          neuromuscular process controlling balance
          detection of mechanical stimulus involved in sensory perception of sound
          regulation of cytosolic calcium ion concentration
          positive regulation of calcium ion transport
          auditory receptor cell stereocilium organization
          inner ear receptor cell differentiation
          calcium ion transmembrane transport
          cochlea development
          calcium ion export
          nucleotide binding
          calcium-transporting ATPase activity
          calcium ion binding
          calmodulin binding
          ATP binding
          protein C-terminus binding
          hydrolase activity
          PDZ domain binding
          calcium-dependent ATPase activity
          acetylcholine receptor binding
          glutamate receptor binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dtm9x:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0