Search
Search

Runx1t1| Gene Symbol: Runx1t1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111026.2 | NP_001104496.1 | 7, 8 | 1411 | ||
| NM_001111027.2 | NP_001104497.1 | 8, 9 | 1605 | |||
| NM_009822.3 | NP_033952.1 | 7, 8 | 1343 | |||
| GenBank mRNA | AB221624.1 | - | 6, 7 | 990 | ||
| AK032132.1 | - | 7, 8 | 1278 | |||
| AK142006.1 | - | 7, 8 | 1412 | |||
| BC139200.1 | - | 7, 8 | 1069 | 8862222 | ||
| BC139201.1 | - | 7, 8 | 1073 | 8862223 | ||
| D32007.1 | - | 7, 8 | 1343 | |||
| X79989.1 | - | 7, 8 | 1050 | |||