Search Thermo Fisher Scientific
Gene Symbol: Serpina6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007618.2 | NP_031644.1 | 3 | 748 | ||
GenBank mRNA | AK145842.1 | - | 3 | 780 | ||
BC013632.1 | - | 3 | 767 | 4194164 | ||
X70533.1 | - | 3 | 748 |