Search
Search

Entpd2| Gene Symbol: Entpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009849.2 | NP_033979.2 | 3 | 385 | ||
| GenBank mRNA | AF042811.1 | - | 3 | 309 | ||
| AK002553.1 | - | 3 | 357 | |||
| AK172281.1 | - | 3 | 333 | |||
| AY376711.1 | - | 3 | 318 | |||
| BC011241.1 | - | 3 | 317 | 3582550 | ||
| BY237099.1 | - | 3 | 386 | |||
| U91511.1 | - | 3 | 352 | |||